C8990
Sp-8-pCPT-cAMPS
≥98% (HPLC), solid
Synonym(s):
8-(4-Chlorophenylthio)adenosine-3′,5′-cyclic monophosphorothioate, Sp-isomer sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H14ClN5O5PS2Na
CAS Number:
Molecular Weight:
509.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥98% (HPLC)
form
solid
packaging
vial of 5 μmol
solubility
H2O: >1 mg/mL
shipped in
dry ice
storage temp.
−70°C
SMILES string
[Na+].Nc1ncnc2n([C@@H]3O[C@@H]4COP([S-])(=O)O[C@H]4[C@H]3O)c(Sc5ccc(Cl)cc5)nc12
General description
Sp-8-pCPT-cAMPS is a nonhydrolyzable analog of cAMP with a p-chlorophenylthio moiety.
Application
Sp-8-pCPT-cAMPS has been used as an agonist for protein kinase A. It has also been used in the inhibition of phosphatidylinositol 4-phosphate 5-kinase (PIP5K) in oligosphere cultures.
Biochem/physiol Actions
Selective cAMP-dependent protein kinase (PKA) activator.
Sp-8-pCPT-cAMPS impairs PIP2 production by phosphatidylinositol 4-phosphate 5-kinase (PIP5K) inhibition. It may be involved in promoting remyelination in astrocytes and neurons. The lipophilicity feature of Sp-8-pCPT-cAMPS aids its cell entry for mediating remyelination. It may play a role in lowering Leishmania proliferation in in vitro studies.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chunyu Zeng et al.
Journal of hypertension, 27(5), 1033-1041 (2009-03-19)
Vascular smooth muscle cell (VSMC) proliferation is central to the development of vascular diseases, including hypertension, which is regulated by numerous hormones and humoral factors. Our previous study showed that the stimulatory effect of norepinephrine on VSMC proliferation is inhibited
Hyun Kyoung Lee et al.
Neuron, 85(6), 1227-1243 (2015-03-11)
Wnt signaling plays an essential role in developmental and regenerative myelination of the CNS; however, contributions of proximal regulators of the Wnt receptor complex to these processes remain undefined. To identify components of the Wnt pathway that regulate these processes
Amrita Saha et al.
Frontiers in pharmacology, 11, 782-782 (2020-07-17)
With the identification of novel cAMP binding effector molecules in Trypanosoma, the role of cAMP in kinetoplastida parasites gained an intriguing breakthrough. Despite earlier demonstrations of the role of cAMP in the survival of Leishmania during macrophage infection, there is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service