Skip to Content
Merck
All Photos(1)

Key Documents

C8759

Sigma-Aldrich

Carisoprodol

Synonym(s):

2-Methyl-2-propyl-1,3-propanediol carbamate isopropylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N2O4
CAS Number:
Molecular Weight:
260.33
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

drug control

USDEA Schedule IV

solubility

ethanol: soluble

originator

Schering Plough

storage temp.

2-8°C

SMILES string

CCCC(C)(COC(N)=O)COC(=O)NC(C)C

InChI

1S/C12H24N2O4/c1-5-6-12(4,7-17-10(13)15)8-18-11(16)14-9(2)3/h9H,5-8H2,1-4H3,(H2,13,15)(H,14,16)

InChI key

OFZCIYFFPZCNJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carisoprodol (N-isopropyl-2-methyl-2-propyl-l,3-propanediol dicarbamate) is a dicarbamate that is structurally similar to mephenesin and meprobamate. It functions as a skeletal muscle relaxant and has analgesic properties. Although it does not have an effect on behavior, it inhibits reticular formation. The area associated with pain perception in the CNS is affected by carisoprodol for its analgesic activity.
Carisoprodol is actively metabolized to meprobamate, a depressant of the CNS having sedative hypnotic.

Application

Carisoprodol has been used as a standard for quantitative determination of carisoprodol in tablets using high-performance thin-layer chromatography. It has also been used to study the effect of pharmacological agents on the muscles of dystrophin-deficient mdx5Cv mice.

Biochem/physiol Actions

Metabolite of meprobamate that is a skeletal muscle relaxant, reduces muscle spasm by depression of brainstem neurons.

Features and Benefits

This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laxmaiah Manchikanti et al.
Pain physician, 16(1), E1-E13 (2013-01-24)
Reports of chronic pain and associated opioid use, abuse, and fatalities continue to increase at an alarming rate, not only in the United States but also across the globe. In light of the many resultant fatalities, multiple authors and authorities
James P Zacny et al.
Drug and alcohol dependence, 120(1-3), 229-232 (2011-08-16)
Some chronic pain patients on long-term opioid therapy also take centrally active skeletal muscle relaxants. One of those muscle relaxants is carisoprodol, a drug that is abused and capable of producing impairment. It would be of relevance to characterize the
Jennifer A Fass
The Annals of pharmacotherapy, 44(12), 1962-1967 (2010-11-11)
To review the current legal status and patterns of abuse of carisoprodol. A literature search was conducted through MEDLINE (1950-August 2010), PubMed (1966-August 2010), EMBASE (1966-August 2010), and International Pharmaceutical Abstracts (1970-August 2010) using the search terms carisoprodol and abuse.
J G Bramness et al.
Clinical pharmacology and therapeutics, 91(3), 438-441 (2012-02-10)
Carisoprodol, a centrally acting muscle relaxant indicated for acute lower back pain, has been available in Europe and the United States since 1959. Studies indicating increased risk of abuse or addiction led to withdrawal of the drug from the market
Michael B Gatch et al.
Drug and alcohol dependence, 123(1-3), 29-34 (2011-11-08)
Carisoprodol is a muscle relaxant that acts at the GABA(A) receptor. Concerns about the abuse liability of carisoprodol are increasing, but evidence that carisoprodol produces tolerance and a significant withdrawal syndrome has yet to be established. The purpose of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service