Skip to Content
Merck
All Photos(1)

Key Documents

C4045

Sigma-Aldrich

Chondroitin disaccharide Δdi-4S sodium salt

≥98% (HPLC)

Synonym(s):

α-ΔUA-[1→3]-GalNAc-4S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H19NO14SNa2
CAS Number:
Molecular Weight:
503.34
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

solubility

H2O: soluble 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].[Na+].CC(=O)N[C@@H](C=O)[C@@H](O[C@@H]1OC(=C[C@H](O)[C@H]1O)C([O-])=O)[C@@H](OS([O-])(=O)=O)[C@H](O)CO

InChI

1S/C14H21NO14S.2Na/c1-5(18)15-6(3-16)11(12(8(20)4-17)29-30(24,25)26)28-14-10(21)7(19)2-9(27-14)13(22)23;;/h2-3,6-8,10-12,14,17,19-21H,4H2,1H3,(H,15,18)(H,22,23)(H,24,25,26);;/q;2*+1/p-2/t6-,7-,8+,10+,11+,12-,14-;;/m0../s1

InChI key

DFRMXDKXNRVAFE-WBOKVGFFSA-L

General description

Chondroitin, a glucosaminoglycan (GAG), is a polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA) that can be sulfated on the either or both GalNAc and GlcA units. Chondroitin and its sulfates are frequently attached to proteins to form proteoglycans.

Application

Chondroitin disaccharide Δdi-4S may be used as a substrate for the identification and characterization of enzymes such as Clostridium perfringens unsaturated glucuronyl hydrolase. Chondroitin disaccharide Δdi-4S may be used as a reference compound in the analysis of chondroitin disaccharide sulfates.

Other Notes

ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GalNAc = N-acetyl-D-galactosamine; 4S = 4-sulfate
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chondroitin sulfate: A key molecule in the brain matrix.
Kwok JC, Warren P, Fawcett JW.
The International Journal of Biochemistry & Cell Biology, 44(4), 582-586 (2011)
Yusuke Nakamichi et al.
The Journal of biological chemistry, 286(8), 6262-6271 (2010-12-15)
Pathogenic Streptococcus agalactiae produces polysaccharide lyases and unsaturated glucuronyl hydrolase (UGL), which are prerequisite for complete degradation of mammalian extracellular matrices, including glycosaminoglycans such as chondroitin and hyaluronan. Unlike the Bacillus enzyme, streptococcal UGLs prefer sulfated glycosaminoglycans. Here, we show
Glucosamine and chondroitin sulfate.
Miller KL, Clegg DO.
Rheumatic Diseases Clinics of North America, 23(1), 103-118 (2011)
Yulin Li et al.
Chemical Society reviews, 41(6), 2193-2221 (2011-11-26)
Injectable hydrogels with biodegradability have in situ formability which in vitro/in vivo allows an effective and homogeneous encapsulation of drugs/cells, and convenient in vivo surgical operation in a minimally invasive way, causing smaller scar size and less pain for patients.
Juliane Salbach et al.
Journal of molecular medicine (Berlin, Germany), 90(6), 625-635 (2011-12-22)
To meet the growing need for tissue replacement materials for our aging population, the development of new adaptive biomaterials is essential. The tissues with the highest demand for implant materials are skin and bone. These tissues share various similarities, including

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service