Skip to Content
Merck
All Photos(1)

Key Documents

C3722

Millipore

C.B.I. supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

wet ice

storage temp.

2-8°C

Application

An antibiotic supplement recommended for the selective isolation of Clostridium botulinum.

Components

Composition: (per vial)
Cycloserine: 125.00 mg
Sulfamethoxazole: 38.00 mg
Trimethoprim: 2.00 mg

Reconstitution

Sufficient for 500 ml medium

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service