C-28041
Pericyte Growth Medium 2
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Growth Medium for Pericytes, Pericyte Medium 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 mL
technique(s)
cell culture | mammalian: suitable
General description
Pericyte Growth Medium 2 (Ready-to-use)
Application
PromoCell Pericyte Growth Medium has been developed for the in vitro cultivation of human pericytes from all tissues. The Medium (Ready-to-use) consists of a 500 mL bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Growth Medium.
Quality
All lots of PromoCell Pericyte Growth Medium are subjected to comprehensive quality control tests using primary human placenta-derived pericytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested pericytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Storage and Stability
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
Storage Class Code
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service