Skip to Content
Merck
All Photos(1)

Key Documents

C237

Sigma-Aldrich

(2S,1′S,2′S)-2-(Carboxycyclopropyl)glycine

solid

Synonym(s):

L-CCG-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO4
CAS Number:
Molecular Weight:
159.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

H2O: >7 mg/mL

SMILES string

N[C@@H]([C@H]1C[C@@H]1C(O)=O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent group II metabotropic glutamate receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer A Feeley Kearney et al.
Experimental neurology, 184 Suppl 1, S30-S36 (2003-11-05)
Metabotropic glutamate receptors (mGluRs) are G-protein-coupled excitatory amino acid (glutamate) receptors and are abundantly expressed in basal ganglia nuclei. We used behavioral, regional glucose uptake metabolic mapping, and FOS protein expression to examine the effects of stimulating striatal and subthalamic
Timo Kirschstein et al.
Neuropharmacology, 47(2), 157-162 (2004-06-30)
Specific agonists of metabotropic glutamate receptors (mGluRs) provide powerful tools to discriminate afferent fibers originating from different presynaptic neurons. The group II mGluR agonists L-CCG-I ((2S,1'S,2'S)-2-(2-carboxycyclopropyl)glycine) and DCG-IV ((2S,2'R,3'R)-2-(2',3'-dicarboxy-cyclopropyl)glycine) are commonly used to distinguish between mossy fiber and associational-commissural (A/C)
Ke-Zhong Shen et al.
The Journal of physiology, 553(Pt 2), 489-496 (2003-09-23)
Patch pipettes were used to record currents in whole-cell configuration to study the effects of group II metabotropic glutamate receptor (mGluR) stimulation on synaptic transmission in slices of rat subthalamic nucleus. Evoked glutamatergic excitatory postsynaptic currents (EPSCs) were reversibly reduced
Cynthia Levinthal et al.
Pharmacology, 83(2), 88-94 (2008-12-10)
Protons act as neuromodulators and produce significant effects on synaptic transmission. The molecular basis of neuromodulation by extracellular protons is partially explained by their effects on certain neurotransmitter receptors and ion channels. The metabotropic glutamate receptors (mGluRs) are a family
Michael Meredith et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(25), 5719-5725 (2004-06-25)
Chemosignals related to reproductive and social status (pheromones) carry messages between opposite-sex and same-sex individuals in many species. Each individual must distinguish signals relevant to its own social behavior with conspecifics from signals used by other (heterospecific) species relevant to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service