Skip to Content
Merck
All Photos(1)

Documents

C-12241

Sigma-Aldrich

Human Pulmonary Artery Endothelial Cells (HPAEC)

500,000 cryopreserved cells

Synonym(s):

Human endothelial cells, Pulmonary artery endothelial cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human pulmonary artery

packaging

pkg of 500,000 cells

morphology

( endothelial)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Artery

Application

Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches) and are provided in a cryopreserved format. Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.

Quality

Rigid quality control tests are performed for each lot of Endothelial Cells from large vessels. They are tested for cell morphology, adherence rate, and cell viability. Flow cytometric analyses for cell-type specific markers, e.g. von Willebrand Factor (vWF) and CD31, as well as Dil-Ac-LDL uptake assays are also carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 5000 - 10000 cells per cm2Passage After Thawing: P2Tested Markers: von Willebrand Factor (vWF) positive, CD31 positive, Dil-Ac-LDL uptake positiveGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Related Content

Cell culture protocol: the endothelial tube formation assay to study angiogenesis using HUVECs and other endothelial cell types. Explore over 350 PromoCell products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service