Skip to Content
Merck
All Photos(2)

Key Documents

BM958.4806.01

B Medical RCW 8

Transport Device (Box)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025

material

polyethylene device

feature

CE compliant: no
CSA approved

packaging

pack of 1 ea

manufacturer/tradename

B Medical Systems

shipped in

ambient

storage temp.

room temp

General description

With 2 ice-packs 0.3 liter + 10 ice-packs 0.6 liter, liquid crystal thermometer, insert small and large), carrying strap, standard packing (0.7 cu ft)

Application

Cold Chain

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service