Skip to Content
Merck
All Photos(1)

Documents

BM0005

Sigma-Aldrich

Avagacestat

≥98% (HPLC)

Synonym(s):

(2R)-2-[N-[(4-Chlorophenyl)sulfonyl]-N-[[2-fluoro-4-(1,2,4-oxadiazol-3-yl)phenyl]methyl]amino]-5,5,5-trifluoropentanamide, (R)-2-(4-Chloro-N-(2-fluoro-4-(1,2,4-oxadiazol-3-yl)benzyl)phenylsulfonamido)-5,5,5-trifluoropentanamide, BMS-708163

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17ClF4N4O4S
CAS Number:
Molecular Weight:
520.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

room temp

SMILES string

NC([C@@H](CCC(F)(F)F)N(S(C1=CC=C(Cl)C=C1)(=O)=O)CC2=C(F)C=C(C3=NOC=N3)C=C2)=O

InChI

1S/C20H17ClF4N4O4S/c21-14-3-5-15(6-4-14)34(31,32)29(17(18(26)30)7-8-20(23,24)25)10-13-2-1-12(9-16(13)22)19-27-11-33-28-19/h1-6,9,11,17H,7-8,10H2,(H2,26,30)/t17-/m1/s1

InChI key

XEAOPVUAMONVLA-QGZVFWFLSA-N

Biochem/physiol Actions

Avagacestat (BMS-708163) is a potent γ-secretase inhibitor selective for amyloid precursor protein (APP) over Notch cleavage. Avagacestat inhibits Aβ40 and Aβ42 production with IC50 values of 0.3 nM and 0.27 nM, respectively, compared to IC50 values of 41, 10, 33, and 38 nM respectively for signaling inhibition of the human Notch 1-4 proteins. At higher concentrations BMS-708163 has been found to reverse sensitivity to the EGFR tyrosine kinase inhibitor gefitinib, thought to be by Notch inhibition.

Features and Benefits

Avagacestat is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service