BAF199230000
Bel-Art® Disposable Pestles
polypropylene, With 1.5 mL Tubes, pack of 100 sets
Synonym(s):
grinding and crushing equipment, plastic pestle, tool for manually grinding solid samples
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polypropylene
packaging
pack of 100 sets
manufacturer/tradename
Bel-Art F19923-0000
L
8.5 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Excellent for resuspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials
- Shaped to optimally grind within 1.5ml microcentrifuge tubes
- BAF199230000 includes 1.5ml microcentrifuge tubes of the same shape
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service