Skip to Content
Merck
All Photos(1)

Documents

B7390

Sigma-Aldrich

Anti-Sheep IgG (whole molecule)–Biotin antibody produced in donkey

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

donkey

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:20,000

storage temp.

2-8°C

Specificity

Binds all sheep Igs

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Gleich et al.
Audiology & neuro-otology, 2(3), 113-127 (1997-05-01)
A monaural conductive hearing loss was induced by interrupting the chain of the middle ear ossicles on the right side in gerbils of four different age groups (P12-14, P20-21, P42 and P84). The volumes of the cochlear nucleus subdivisions and
Niki Kourmouli et al.
Journal of cell science, 117(Pt 12), 2491-2501 (2004-05-07)
Tri-methylated lysine 20 on histone H4 (Me(3)K20H4) is a marker of constitutive heterochromatin in murine interphase and metaphase cells. Heterochromatin marked by Me(3)K20H4 replicates late during S phase of the cell cycle. Serum starvation increases the number of cells that
J H Y Yeung et al.
IBRO neuroscience reports, 13, 78-86 (2023-01-03)
Glutamate is the main excitatory neurotransmitter in the central nervous system, responsible for a plethora of cellular processes including memory formation and higher cerebral function and has been implicated in various neurological disease states. Alzheimer's disease (AD) is the leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service