Skip to Content
Merck
All Photos(1)

Key Documents

B5559

Sigma-Aldrich

BRL 52537 hydrochloride

>98% (HPLC)

Synonym(s):

(±)-1-(3,4-Dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2OCl2 · HCl
Molecular Weight:
391.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

color

off-white

solubility

methanol: soluble ~18 mg/mL at 60 °C
DMSO: insoluble
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cl[H].Clc1ccc(CC(=O)N2CCCCC2CN3CCCC3)cc1Cl

Gene Information

Biochem/physiol Actions

κ/μ opioid receptor agonist.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service