Skip to Content
Merck
All Photos(3)

Key Documents

AV32797

Sigma-Aldrich

Anti-SCD antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Stearoyl-CoA desaturase (δ-9-desaturase)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCD(6319)

General description

Rabbit polyclonal anti-SCD antibody reacts with human and bovine stearoyl-CoA desaturases.
Stearoyl-CoA desaturase (SCD) is a rate-limiting enzyme that catalyses the conversion of saturated fatty acids to monounsaturated fatty acids (MUFA). SCD functions as a homeostatic check-point between glucose and fatty acid metabolism. It is a key enzyme target in the search for potential drugs to manage obesity.

Immunogen

Synthetic peptide directed towards the middle region of human SCD

Application

Rabbit Anti-SCD antibody can be used for western blot applications at a concentration of 1.0 μg/ml. It can also be used for IHC at 4-8 μg/ml, using paraffin-embedded tissues.
Rabbit polyclonal anti-SCD antibody is used to tag stearoyl-CoA desaturase for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of stearoyl-CoA desaturases in the homeostasis of fatty acid metabolism at the level of monounsaturation.

Biochem/physiol Actions

Stearoyl-CoA desaturase ( fatty acid desaturase, SCD) is expressed at high levels in several human tissues and is required for the biosynthesis of oleate (18:1) and palmitoleate (16:1). These monounsaturated fatty acids are the major components of phospholipids, triglycerides, wax esters, and cholesterol esters.

Sequence

Synthetic peptide located within the following region: HPAVKEKGSTLDLSDLEAEKLVMFQRRYYKPGLLMMCFILPTLVPWYFWG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana S H Costa et al.
PloS one, 13(4), e0193875-e0193875 (2018-04-04)
Despite the recent advances in transcriptomics, gene expression studies addressing cattle´s skeletal muscle adaptations in response to compensatory growth are warranted, particularly regarding lipid metabolism due to its impact in meat sensory and nutritional traits. In the present study, in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service