Skip to Content
Merck
All Photos(2)

Documents

AV02065

Sigma-Aldrich

Anti-TNFRSF10A antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Tumor necrosis factor receptor superfamily, member 10a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

50 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

The immunogen for anti-TNFRSF10A antibody: synthetic peptide derected towards the C terminal of human TNFRSF10A

Application

Anti-TNFRSF10A antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

TNFRSF10A or Death receptor 4 is a mediator of apoptosis and tumor suppression. Mutation in TNFRSF10A gene or dysfunction of the receptor results in tumor development and invasion. Polymorphism of Glu228Ala residue in TNFRSF10A is identified as a risk factor in metastatic prostate cancer after radiation therapy.

Sequence

Synthetic peptide located within the following region: RAGTAGPGDALYAMLMKWVNKTGRNASIHTLLDALERMEERHAKEKIQDL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanja Langsenlehner et al.
The Prostate, 68(3), 264-268 (2008-01-01)
Death receptor 4, encoded by the TNFRSF10A gene, is an important mediator of apoptosis and its dysfunction may be related to cancer development and distant tumor spread. A single nucleotide polymorphism in TNFRSF10A (Glu228Ala, rs20576) within a conserved region of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service