Skip to Content
Merck
All Photos(1)

Documents

A8892

Sigma-Aldrich

Allyl bromide

Synonym(s):

3-Bromo-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2Br
CAS Number:
Molecular Weight:
120.98
Beilstein:
605308
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

4.2 (vs air)

autoignition temp.

554 °F

expl. lim.

7.3 %

refractive index

n20/D 1.469 (lit.)

bp

70-71 °C (lit.)

mp

−119 °C (lit.)

density

1.398 g/mL at 25 °C (lit.)

SMILES string

BrCC=C

InChI

1S/C3H5Br/c1-2-3-4/h2H,1,3H2

InChI key

BHELZAPQIKSEDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service