Skip to Content
Merck
All Photos(1)

Key Documents

A8099

Spectrafuge 7M, 16M and Minifuge Adapters

0.5 mL tubes

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€52.70

€52.70


Estimated to ship onApril 29, 2025Details



Select a Size

Change View
1 EA
€52.70

About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

€52.70


Estimated to ship onApril 29, 2025Details


manufacturer/tradename

Labnet

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Package Qty: Adapters supplied as a set of 6

Legal Information

Spectrafuge is a trademark of Labnet Intl., Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service