Skip to Content
Merck
All Photos(1)

Key Documents

A5949

Sigma-Aldrich

L-Arginine monohydrochloride

BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), >99% (TLC)

Synonym(s):

(S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
EC Number:
MDL number:
UNSPSC Code:
12352209

product line

BioXtra

Assay

>99% (TLC)

impurities

<0.0005% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.1%

pH

5.0-7.0 (20 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%

absorption

<0.1 at 280 in H2O at 1 M
<0.2 at 260 in H2O at 1 M

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Di Somma et al.
Endocrine, 49(2), 492-502 (2014-11-10)
The aim of the study is to clarify the relationship between adipose tissue dysfunction, metabolic profile and growth hormone (GH)/insulin-like growth factor (IGF)-I secretion in healthy adult subjects. We investigated the metabolic profile in a cohort of 231 consecutive healthy
C B Christiansen et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 47(7), 537-543 (2015-01-21)
TLQP-21 is a VGF-derived neuropeptide proposed to be involved in regulation of metabolism. More specifically it has been suggested that TLQP-21 has the ability to enhance glucose stimulated insulin secretion, making it a candidate for treatment of patients with type
N E Paul et al.
Bio-medical materials and engineering, 25(3), 289-297 (2015-09-26)
Adipose tissue engineering is a promising solution for the reconstruction of soft tissue defects. An insufficient neovascularisation within the scaffolds that leads to necrosis and tissue loss is still a major shortcoming of current tissue engineering attempts. Biomaterials, which release
Nivaldo Ribeiro Villela et al.
Journal of translational medicine, 12, 232-232 (2014-08-26)
Relative hypovolemia is frequently found in early stages of severe sepsis and septic shock and prompt and aggressive fluid therapy has become standard of care improving tissue perfusion and patient outcome. This paper investigates the role of the nitric oxide
Jenna Gallie et al.
PLoS biology, 13(3), e1002109-e1002109 (2015-03-13)
Phenotype switching is commonly observed in nature. This prevalence has allowed the elucidation of a number of underlying molecular mechanisms. However, little is known about how phenotypic switches arise and function in their early evolutionary stages. The first opportunity to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service