Skip to Content
Merck
All Photos(1)

Key Documents

A5727

Sigma-Aldrich

N-[(4S)-4-amino-5-[(2-aminoethyl)amino]pentyl]-N′-nitroguanidine tris(trifluoroacetate) salt

≥98% (HPLC), powder

Synonym(s):

N-(4S)-(4-amino-5-[aminoethyl]aminopentyl)-N′-nitroguanidine tris(trifluoroacetate) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24N7O8F9
CAS Number:
Molecular Weight:
589.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

solubility

H2O: soluble 50 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.NCCNC[C@@H](N)CCCNC(=N)N[N+]([O-])=O

InChI

1S/C8H21N7O2.3C2HF3O2/c9-3-5-12-6-7(10)2-1-4-13-8(11)14-15(16)17;3*3-2(4,5)1(6)7/h7,12H,1-6,9-10H2,(H3,11,13,14);3*(H,6,7)/t7-;;;/m0.../s1

InChI key

NYWNMNBIORWOSQ-QTPLPEIMSA-N

Biochem/physiol Actions

Potent and selective inhibitor of nNOS (neuronal nitric oxide synthase).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Marie Pelletier et al.
American journal of physiology. Gastrointestinal and liver physiology, 298(6), G896-G907 (2010-03-27)
Hyperplasia of smooth muscle contributes to the thickening of the intestinal wall that is characteristic of inflammation, but the mechanisms of growth control are unknown. Nitric oxide (NO) from enteric neurons expressing neuronal NO synthase (nNOS) might normally inhibit intestinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service