Skip to Content
Merck
All Photos(1)

Key Documents

94206

Sigma-Aldrich

Undecyl β-D-maltoside

≥99.0% (TLC)

Synonym(s):

Undecyl β-D-maltopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€400.00

€400.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€400.00

About This Item

Empirical Formula (Hill Notation):
C23H44O11
CAS Number:
Molecular Weight:
496.59
Beilstein:
9372606
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

€400.00


Please contact Customer Service for Availability

Request a Bulk Order

description

non-ionic

Assay

≥99.0% (TLC)

form

powder

optical activity

[α]20/D +49.0±2.0°, c = 2% in H2O

mol wt

496.59 g/mol

CMC

0.59 mM (20-25°C)

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OCCCCCCCCCCC)O[C@@H]2CO)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C23H44O11/c1-2-3-4-5-6-7-8-9-10-11-31-22-20(30)18(28)21(15(13-25)33-22)34-23-19(29)17(27)16(26)14(12-24)32-23/h14-30H,2-13H2,1H3/t14-,15-,16-,17+,18-,19-,20-,21-,22-,23-/m1/s1

InChI key

UYEMNFYVTFDKRG-ZNGNCRBCSA-N

Application

Detergent for the purification, extraction and solubilization of membrane-bound proteins in the native structure. It increases solubilization by dissociating aggregates and is easily removed by dialysis.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ostermeier, C., et al.
Proceedings of the National Academy of Sciences of the USA, 94, 105476-105476 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service