Skip to Content
Merck
All Photos(1)

Key Documents

93015

Sigma-Aldrich

TRIS Glycine buffer solution

BioReagent, for molecular biology, 10x concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 L
€537.00

€537.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 L
€537.00

About This Item

EC Number:
UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25

€537.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

liquid

quality

10x concentrate

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

8.6±0.2

density

1.06 g/mL at 20 °C

application(s)

diagnostic assay manufacturing

Looking for similar products? Visit Product Comparison Guide

Physical form

0.25 M TRIS, 1.92 M glycine

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service