Skip to Content
Merck
All Photos(1)

Key Documents

92689

Sigma-Aldrich

N-Acetyl-D-galactosamine-6-phosphate

≥97.0% (HPLC)

Synonym(s):

N-Acetyl-D-chondrosamine 6-phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€104.00

€104.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€104.00

About This Item

Empirical Formula (Hill Notation):
C8H16NO9P
CAS Number:
Molecular Weight:
301.19
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

€104.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.0% (HPLC)

form

crystalline

color

light yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](COP(O)(O)=O)[C@H](O)[C@@H]1O

InChI

1S/C8H16NO9P/c1-3(10)9-5-7(12)6(11)4(18-8(5)13)2-17-19(14,15)16/h4-8,11-13H,2H2,1H3,(H,9,10)(H2,14,15,16)/t4-,5-,6+,7-,8?/m1/s1

InChI key

BRGMHAYQAZFZDJ-KEWYIRBNSA-N

Biochem/physiol Actions

N-Acetyl-D-galactosamine-6-phosphate has been shown to inhibit the haemagglutinating activity of Alectin II, a compound isolated from aloe vera leaf pulp. [1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Akev et al.
Phytotherapy research : PTR, 13(6), 489-493 (1999-09-10)
The separation and partial purification of two lectins from the leaf pulp of Aloe vera L. (=barbadensis Miller) is presented. The fraction showing haemagglutinating activity was precipitated at 50% ammonium sulphate concentration from the crude leaf pulp extract. The precipitate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service