Skip to Content
Merck
All Photos(1)

Key Documents

89380

Sigma-Aldrich

Deoxyribonucleic acid sodium salt from calf thymus

lyophilized

Synonym(s):

DNA sodium salt from calf thymus, Thymonucleic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106310
eCl@ss:
32160414
NACRES:
NA.52
Product 89380 is not currently sold in your country. Contact Technical Service

grade

for molecular biology

Quality Level

form

lyophilized

quality

lyophilized

impurities

salt, none detected
≤5% protein (BCA)

absorbance ratio

A260/280 nm ~1.6

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Deoxyribonucleic acid sodium salt from calf thymus was used in the preparation of DNA-binding protein-sized quantum dots.[1]

Quantity

A260/mg >13,

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temperature-and salt-dependent binding of long DNA to protein-sized quantum dots: thermodynamics of ?inorganic protein?-DNA interactions
Rahina M et al
Journal of the American Chemical Society, 122, 14-17 (2000)
Youngsil Seo et al.
Scientific reports, 10(1), 2140-2140 (2020-02-09)
Intrabodies, antibodies expressed within cells, offer an interesting way to target intracellular molecules, making them potentially useful for biotechnology and medicine. However, it remains controversial whether full-size IgG intrabodies expressed in the reducing environment of the cytosol of mammalian cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service