The estimated molecular weight range of this product is 50,000 and 100,000 Da which is approximately 75-150 base pairs. The product is a double-strand DNA.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 g | Please contact Customer Service for Availability | €63.80 |
100 g | Please contact Customer Service for Availability | €433.00 €368.05 |
About This Item
CAS Number:
UNSPSC Code:
41106310
eCl@ss:
32160414
EC Number:
309-566-6
NACRES:
NA.31
MDL number:
Form:
powder
Storage temp.:
2-8°C
€63.80
Please contact Customer Service for Availability
form
powder
Quality Segment
impurities
≤1% protein
loss
≤8% loss on drying
absorbance ratio
A260/280 nm ≥1.5
storage temp.
2-8°C
Application
Deoxyribonucleic acid from fish sperm has been used:
- to study the effect of DNA in Maillard reaction.
- as a standard for DNA measurements.
- asa component of artificial sputum media (ASM), which is designed to simulate theconditions within the cystic fibrosis (CF) airway for growing bacterialcultures.
Other Notes
Not available for import to United States
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 200 | Quality Level - | Quality Level 200 |
| form powder | form solid | form lyophilized powder | form powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| impurities ≤1% protein | impurities - | impurities ≤5% protein | impurities - |
| absorbance ratio A260/280 nm ≥1.5 | absorbance ratio - | absorbance ratio A260/280 nm ~1.4 | absorbance ratio - |
| loss ≤8% loss on drying | loss - | loss - | loss - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Could you please inform us whether the nucleic acid is double-stranded or single-stranded? Additionally, we would appreciate an estimate of its length in base pairs, even if approximate.
1 answer-
Helpful?
-



