58955C
50% Glucose (w/v) – HTST Treated
sterile-filtered, animal-component free solution
Pharma Manufacturing
About This Item
Recommended Products
description
For Research or Further Manufacturing Uses Only - Not Intended for Direct Use In Humans or Animals
HTST treated
Quality Level
sterility
sterile-filtered
form
liquid
quality
endotoxin tested
shelf life
24 mo.
packaging
bag of (single use)
shipped in
ambient
storage temp.
room temp
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service