Skip to Content
Merck
All Photos(1)

Key Documents

4390731

Sigma-Aldrich

iTRAQ® Reagent - 8PLEX 50U Bulk Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
Pricing and availability is not currently available.

storage temp.

−20°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

General description

Includes 8 tubes. Each tube contains 50 units of an individual 8-plex reagent. This product contains the reagents in the equivalent Sciex iChemistry® product 4393529.

Analysis Note

To view the Protocol for the iTRAQ Reagents 8-plex Kit, please visit this Protocol link.

To view the Chemistry Quick Reference Card for the iTRAQ Reagents, please visit this Chemistry Quick Reference Card link.

Legal Information

iChemistry is a registered trademark of AB Sciex Pte. Ltd.
iTraq is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service