Skip to Content
Merck
All Photos(1)

Key Documents

36297

Sigma-Aldrich

Bimane amine

BioReagent, suitable for fluorescence, ≥98.0% (HPCE)

Synonym(s):

3-Aminomethyl-2,5,6-trimethyl-1H,7H-pyrazolo[1,2-a]pyrazole-1,7-dione, Aminobimane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O2
CAS Number:
Molecular Weight:
207.23
Beilstein:
4186574
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥98.0% (HPCE)

form

solid

fluorescence

λex 386 nm; λem 480 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC1=C(C)C(=O)N2N1C(CN)=C(C)C2=O

InChI

1S/C10H13N3O2/c1-5-7(3)12-8(4-11)6(2)10(15)13(12)9(5)14/h4,11H2,1-3H3

InChI key

WLADQYKRYSPMME-UHFFFAOYSA-N

Application

Bimane amine (Aminobimane) may be useful as a fluorescent probe for molecules that react with primary amines.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service