Skip to Content
Merck
All Photos(1)

Key Documents

3445-001-01

Cultrex® 3-D Culture Matrix Reduced Growth Factor Basement Membrane Extract, PathClear®

size 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

concentration

14-16 mg/mL

size

1 mL

shipped in

dry ice

storage temp.

−70°C

General description

3D Culture is a cell culture method that provides cells with the necessary structure and signaling cues to direct reconstruction of the tissue architecture, and these methods provide physiologically predictive in vitro models for evaluating development and disease. Normal cells assemble into organiods that structurally resemble their emanating tissues, exhibit a polarized morphology, undergo cell cycle regulation, and produce tissue-specific proteins. While cancer cells assemble into tumor-like structures, lacking an organized architecture or cell cycle regulation and exhibiting tumor-specific markers depending on the extend of malignancy.

Physical form

Solution in Dulbecco′s Modified Eagle′s medium without phenol red, with 10 ug/ml gentamicin sulfate.

Other Notes

Functional Assay:
Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells.
3D Culture - Basement Membrane Extract promotes differentiation of a human epithelial cell line derived from mammary gland (MCF-10A) or human prostate (PC-3) into acinar structures.
Product is stable for a minimum of 3 months from date of shipment when stored at –20°C in a manual defrost freezer. For optimal stability, store at –80°C. Avoid freeze-thaw cycles.
Sterility Testing:
PathClear® - Negative by PCR test for mycoplasma; 17 bacterial and virus strains typically included in mouse antibody production (MAP) testing, plus 13 additional murine infectious agents including LDEV, for a total of 31 organisms and viruses. No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP sterility testing guidelines. Endotoxin concentration = 8 EU/ml by LAL assay.

Legal Information

3-D Culture Matrix is a trademark of Trevigen, Inc.
Cultrex is a registered trademark of Trevigen, Inc.
PathClear is a registered trademark of Trevigen, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service