All Photos(1)
About This Item
Linear Formula:
H2NC6H4CONH2
CAS Number:
Molecular Weight:
136.15
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
97%
mp
115-116 °C (lit.)
SMILES string
NC(=O)c1cccc(N)c1
InChI
1S/C7H8N2O/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H2,9,10)
InChI key
GSCPDZHWVNUUFI-UHFFFAOYSA-N
Gene Information
mouse ... Parp1(11545)
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Li-qun Zhang et al.
Shock (Augusta, Ga.), 37(5), 492-500 (2012-01-24)
The overactivation of the nuclear enzyme poly(ADP-ribose) polymerase (PARP) is considered a final common effector in ischemia/reperfusion (I/R) injury. The aim of the current study was to examine the precise time course of the activation of PARP in peripheral leukocytes
Wen Ji Li et al.
The journal of sexual medicine, 9(5), 1319-1327 (2012-03-21)
Endothelial dysfunction-induced abnormalities of the nitric oxide (NO)/cyclic guanosine monophosphate (cGMP) signaling pathway in the corpus cavernosum are thought to be the main factors involved in the pathogenesis of diabetes-induced erectile dysfunction (ED). Recent studies have shown that the poly(adenosine
Shunichi Shimizu et al.
The international journal of biochemistry & cell biology, 68, 119-127 (2015-09-20)
Transient receptor potential melastatin 2 (TRPM2) is an oxidative stress-sensitive Ca(2+)-permeable channel. In monocytes/macrophages, H2O2-induced TRPM2 activation causes cell death and/or production of chemokines that aggravate inflammatory diseases. However, relatively high concentrations of H2O2 are required for activation of TRPM2
Dan Huang et al.
PloS one, 6(10), e27123-e27123 (2011-11-11)
Transforming growth factor type-β (TGF-β)/Smad pathway plays an essential role in vascular fibrosis. Reactive oxygen species (ROS) generation also mediates TGF-β signaling-induced vascular fibrosis, suggesting that some sort of interaction exists between Smad and redox pathways. However, the underlying molecular
Maria I Behrens et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 67(10), 1036-1040 (2012-03-01)
A paucity of cancer in individuals with Alzheimer's disease (AD) and low rates of AD in cancer survivors has been reported in epidemiological studies. Deregulation in opposite directions of biological mechanisms, such as susceptibility to cell death, might be shared
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service