Skip to Content
Merck
All Photos(1)

Documents

19773

Sigma-Aldrich

Nε,Nε-Dimethyl-L-lysine monohydrochloride

≥96% (TLC)

Synonym(s):

(S)-2-Amino-6-(dimethylamino)hexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O2 · HCl
CAS Number:
Molecular Weight:
210.70
Beilstein:
5992298
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.28

product name

Nε,Nε-Dimethyl-L-lysine monohydrochloride, ≥96% (TLC)

Assay

≥96% (TLC)

form

powder

optical activity

[α]/D +11.5±1.0°, c = 1 in H2O

color

white

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCCC[C@H](N)C(O)=O

InChI

1S/C8H18N2O2.ClH/c1-10(2)6-4-3-5-7(9)8(11)12;/h7H,3-6,9H2,1-2H3,(H,11,12);1H/t7-;/m0./s1

InChI key

CRNNIVHOUZQLSF-FJXQXJEOSA-N

Biochem/physiol Actions

Metabolite found in various sources, like human urine, Salmonella typhimurium flagella,and amoeba actin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of episilon-N-methyllysine in Spirillum serpens flagella and of episilon-N-dimethyllysine in Salmonella typhimurium flagella.
A N Glazer et al.
Biochimica et biophysica acta, 188(1), 164-165 (1969-08-12)
Isolation and identification of N-G,N-G- and N-G,N'-G-dimethyl-arginine, N-epsilon-mono-, di-, and trimethyllysine, and glucosylgalactosyl- and galactosyl-delta-hydroxylysine from human urine.
Y Kakimoto et al.
The Journal of biological chemistry, 245(21), 5751-5758 (1970-11-10)
Epsilon-N-dimethyllysine in amoeba actin.
R R Weihing et al.
Nature, 227(5264), 1263-1264 (1970-09-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service