Skip to Content
Merck

18521

6-Fmoc-amino-D-luciferin

≥98.5% (TLC)

Synonym(s):

(4S)-2-[6-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-2-benzothiazolyl]-4,5-dihydro-4-thiazolecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H19N3O4S2
CAS Number:
Molecular Weight:
501.58
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.5% (TLC)

form

solid

solubility

DMSO: 1 mg/mL, clear, colorless to almost colorless

functional group

Fmoc

storage temp.

−20°C

SMILES string

O=C(NC1=CC(SC(C2=N[C@](C(O)=O)([H])CS2)=N3)=C3C=C1)OCC4C(C=CC=C5)=C5C6=C4C=CC=C6

InChI

1S/C26H19N3O4S2/c30-25(31)21-13-34-23(29-21)24-28-20-10-9-14(11-22(20)35-24)27-26(32)33-12-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-11,19,21H,12-13H2,(H,27,32)(H,30,31)/t21-/m1/s1

InChI key

ZKXGWUCUGSIASJ-OAQYLSRUSA-N

General description

Amino analog of D-luciferin substrate for firefly luciferase activity. Useful in automated peptide synthesis for incorporation of amino-luciferin into synthetic peptides.

Application

Peptides containing an aminoluciferin become sensitive chemiluminescent substrates for their individual specific peptidase activity when luciferase is added.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library