Skip to Content
Merck
All Photos(1)

Key Documents

05881

Sigma-Aldrich

3-Amino-5-hydroxybenzoic acid hydrochloride

≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO3 · HCl
CAS Number:
Molecular Weight:
189.60
Beilstein:
5305404
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0% (HPLC)
97.0-103.0% (AT)

form

powder

mp

≥300 °C

SMILES string

Cl.Nc1cc(O)cc(c1)C(O)=O

InChI

1S/C7H7NO3.ClH/c8-5-1-4(7(10)11)2-6(9)3-5;/h1-3,9H,8H2,(H,10,11);1H

InChI key

CXESTILCPSBCGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite in the amino-shikimate pathway[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiantao Guo et al.
Journal of the American Chemical Society, 124(4), 528-529 (2002-01-24)
With respect to the source of the nitrogen atom incorporated into the aminoshikimate pathway, d-erythrose 4-phosphate has been proposed to undergo a transamination reaction resulting in formation of 1-deoxy-1-imino-d-erythrose 4-phosphate. Condensation of this metabolite with phosphoenolpyruvate catalyzed by aminoDAHP synthase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service