Skip to Content
Merck
All Photos(1)

Key Documents

W584649

Sigma-Aldrich

Cocoa extract

synthetic blend, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12164502
grade:
FG
Halal
Kosher
food allergen:
no known allergens

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 182.20

form

liquid

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cocoa

Application

Ingredients Declaration
Ingredient: Natural Cocoa Extract > 65%; Flavoring Substances > 20%; Caramel E150a > 10% (color); MPG > 6% (carrier); Water < 0.5% (carrier); Ethanol <0.004%(carrier)

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service