Skip to Content
Merck
All Photos(1)

Documents

W1386

Millipore

Wilson Blair agar base

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607

application(s)

food and beverages
microbiology

suitability

Salmonella spp.

Application

Recommended for the isolation of Salmonellae.

Components

Ingredients (g/L):
Peptone, special, 10.00
Beef extract, 5.00
Dextrose, 10.00
Sodium chloride, 5.00
Agar, 30.00

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service