Skip to Content
Merck
All Photos(1)

Documents

SK016

Supelco

HVOC CALIBRATION VERIFICATION

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

Quality Level

format

multi-component solution

storage temp.

15-25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    1,1,1-Trichloroethane 20.0 µg/mL
    1,1,2-Trichloro-1,2,2-trifluoroethane 20.0 µg/mL
    1,1,2-Trichloroethane 20.0 µg/mL
    1,2,3-Trichlorobenzene 20.0 µg/mL
    1,2,4-Trichlorobenzene 20.2 µg/mL
    1,2-Dichlorobenzene 20.0 µg/mL
    1,3,5-Trichlorobenzene 20.0 µg/mL
    1,3-Dichlorobenzene 20.3 µg/mL
    1,4-Bromochlorobenzene 20.0 µg/mL
    1,4-Dichlorobenzene 20.0 µg/mL
    Carbon Tetrachloride 10.5 µg/mL
    Methylene Chloride 49.9 µg/mL
    Tetrachloroethylene 20.3 µg/mL
    Trichloroethene 20.0 µg/mL
    Trichlorofluoromethane 10.1 µg/mL
    See All (15)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-11.99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service