Skip to Content
Merck
All Photos(1)

Documents

PHR8667

Supelco

Oligonucleotide HPLC Performance Standard Mix, 12-33 NT

new

Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

grade

pharmaceutical secondary standard
reference material

Quality Level

200
300

CofA

current certificate can be downloaded

packaging

pkg of 0.8 - 1 nmol (per tube, each analyte)

application(s)

pharmaceutical

storage temp.

-10 to -25°C

General description

The Oligonucleotide HPLC Performance Standard Mix (Oligo-6) is a calibration standard with carefully selected oligonucleotides to test and monitor changes in HPLC and LC/MS Systems. The difference in the selectivity of the oligonucleotides make it useful for most chromatographic modes including Ion Pairing Reversed Phase, Ion Exchange and HILIC Methods.

The oligonucleotides in the mix are dried and between 12-33 NT that makes it a suitable tool alongside development and quality control of therapeutics like antisense oligonucleotides, small interfering RNAs, and aptamers.

Composition

Sequence 1, 12 NT, 0.8 nmol, MW 3588.4

Sequence 2, 12 NT, 1.0 nmol, MW 4158.0

Sequence 3, 25 NT, 1.0 nmol, MW 7580.9

Sequence 4, 33 NT, 1.0 nmol, MW 10014.4

Sequence 5, 20 NT, 1.0 nmol, MW 6117.0

Sequence 6, 15 NT, 1.0 nmol, MW 4395.9

Application

This Oligonucleotide Standard Mix are used for:

  • System suitability and creation of control charts
  • Method transfer and trouble shooting
  • Monitoring of HPLC Column life span IPRP-HPLC, IEX HPLC, HILIC, LCMS

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service