Skip to Content
Merck
All Photos(1)

Key Documents

PHR3689

Telmisartan Related Compound B

new

pharmaceutical secondary standard, certified reference material

Synonym(s):

4′-[(1,7′-Dimethyl-2′-propyl[2,5′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, Telmisartan Related Compound B, 4′-[(1,7′-Dimethyl-2′-propyl-1H,1′H-2,5′-bibenzo[d]imidazol-1′-yl)methyl]biphenyl-2-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

PHR3689-40MG
€361.00

€361.00


Please contact Customer Service for Availability


Select a Size

Change View
PHR3689-40MG
€361.00

About This Item

Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
UNSPSC Code:
12352200

€361.00


Please contact Customer Service for Availability

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1643437

API family

telmisartan

packaging

pkg of 40MG

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

[n]3(c4c(nc3CCC)cc(cc4C)c5[n](c6c(n5)cccc6)C)Cc1ccc(cc1)c2c(cccc2)C(=O)O

InChI

1S/C33H30N4O2/c1-4-9-30-34-28-19-24(32-35-27-12-7-8-13-29(27)36(32)3)18-21(2)31(28)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)

InChI key

COYNILPBALLLCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service