PHR3666
Ipratropium Bromide Related Compound B

pharmaceutical secondary standard
Synonym(s):
(1R,3r,5S,8s)-3-[[(2RS)-3-Hydroxy-2-phenylpropanoyl]oxy]-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane bromide
Select a Size
€366.00
Select a Size
About This Item
€366.00
Quality Level
grade
pharmaceutical secondary standard
Agency
traceable to EP I0361000
traceable to USP 1347529
API family
ipratropium
CofA
current certificate can be downloaded
packaging
pkg of 30 mg
application(s)
pharmaceutical
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service