PHR2469
Guaiacol Impurity C
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether, Veratrole
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
guaiacol
form
liquid
packaging
pkg of 100 mg
refractive index
n20/D 1.533 (lit.)
bp
206-207 °C (lit.)
mp
15 °C (lit.)
density
1.084 g/mL at 25 °C (lit.)
application(s)
pharmaceutical small molecule
storage temp.
2-30°C
SMILES string
COc1ccccc1OC
InChI
1S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6H,1-2H3
InChI key
ABDKAPXRBAPSQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service