NIST764A
Magnetic susceptibility standard - platinum cylinder
Select a Size
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 3 mm dia. x 3.42 L
manufacturer/tradename
NIST®
application(s)
general analytical
Related Categories
General description
SRM 764A_cert
SRM 764A_SDS
Other Notes
Platinum (Pt)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service