Skip to Content
Merck
All Photos(1)

Key Documents

NIST617

Trace elements in glass

NIST SRM 617

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available. Contact Technical Service

grade

certified reference material

Quality Level

packaging

pkg of 4 wafers

manufacturer/tradename

NIST®

application(s)

glass & ceramic

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Certified for the analytes listed below.
Lead (Pb), Potassium (K), Strontium (Sr), Thorium (Th), Uranium (U)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Boron (B), Copper (Cu), Gallium (Ga), Gold (Au), Iron (Fe), Lanthanum (La), Rubidium (Rb), Scandium (Sc), Thallium (Th), Titanium (Ti), Strontium (87Sr/86Sr), Uranium (235U/UTotal)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service