Skip to Content
Merck
All Photos(1)

Key Documents

NIST2165

Low alloy steel

NIST® SRM® 2165

Sign Into View Organizational & Contract Pricing

Select a Size

150 G
€1,280.00

€1,280.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
150 G
€1,280.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,280.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 150 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is low alloy steel in the form of chips sized to pass through sieve openings between 0.50 mm and 1.18 mm (35 mesh to 16 mesh). It is intended primarily for use in evaluation of chemical and instrumental methods of analysis of steel and materials of similar matrix. It should not be used for calibration. A unit of SRM 2165 consists of one bottle containing approximately 150 g of chips. For more information, please refer to the COA and SDS.

SRM 2165_cert

SRM 2165_SDS

Other Notes

Certified for the analytes listed below.
Antimony (Sb), Arsenic (As), Chromium (Cr), Cobalt (Co), Copper (Cu), Lead (Pb), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Niobium (Nb), Phosphorus (P), Silver (Ag) Sulfur (S), Tin (Sn), Titanium (Ti), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Boron (B), Carbon (C), Aluminum (Al), Bismuth (Bi), Magnesium (Mg), Selenium (Se), Silicon (Si), Tantalum (Ta), Tellurium (Te)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service