Skip to Content
Merck
All Photos(1)

Key Documents

HLA001

Sigma-Aldrich

Raji HLA Panel Cell Lines

human lymph, lymphoblast

Synonym(s):

Raji cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Product HLA001 is not currently sold in your country. Contact Technical Service

Product Name

Raji HLA Panel Cell Lines,

biological source

human lymph

growth mode

suspension

morphology

lymphoblast

shipped in

dry ice

storage temp.

−196°C

General description

The STR profile of these cell lines match that of its parental cell line - ECACC catalog number 85011429.

This product is a panel of nine (9) genetically modified cell lines targeting MHC Class I HLA molecules in a Raji lymphoblast parental cell line background.  The full HLA panel consists of a Beta-2 microglobulin (β2M) knockout cell line and an additional eight (8) monoallelic HLA expression cell lines.  

CompoZr® Zinc Finger Nuclease (ZFN) technology was used to generate the β2M knockout cell line, which does not express endogenous cell surface MHC Class I HLA molecules (HLA-A or HLA-B) as measured by fluorescence-activated cell sorting (FACS).  

MISSION® lentivirus was used to generate the eight (8) monoallelic HLA cell lines, which express individual HLA-A or HLA-B subtypes on the cell surface via a β2M:HLA fusion protein as described in Nature Biotechnology 35, 765-772, (2017) and as demonstrated via FACS analysis.    

Cell Line Description

Established in 1963 from Burkitt′s lymphoma in an 11 year old male. Growth is in the form of single cells without attachment and as macroscopically visible clumps containing many hundreds of cells. Resistant to VSV. This cell line carries the latent Epstein-Barr Virus (EBV) genome and is positive for EBNA. Raji is sometimes referred to as a ′non-producer′; the EBV genome carries deletions attributed to preventing the formation of virus particles. Ethnicity: Black

Features and Benefits

Unlike wild type Raji cells, which express multiple HLA subtypes, the Raji HLA panel cell lines express individual MHC Class I HLA subtypes. These monoallelic HLA cell lines can be used to study peptide binding and presentation in the context of individual HLA subtypes. The panel can also be used to develop cell-based assays to evaluate candidate cancer immunotherapies and identify tumor-specific neoantigens.

Components

HLA001A-1VL: Raji B2M KO Cells
HLA001B-1VL: Raji HLA-A*02:01 Cells
HLA001C-1VL: Raji HLA-A*01:01 Cells
HLA001D-1VL: Raji HLA-A*03:01 Cells
HLA001E-1VL: Raji HLA-A*11:01 Cells
HLA001F-1VL: Raji HLA-A*24:02 Cells
HLA001G-1VL: Raji HLA-B*15:10 Cells
HLA001H-1VL: Raji HLA-B*07:02 Cells
HLA001K-1VL: Raji HLA-B*40:01 Cells

Quality

Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification.

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service