Skip to Content
Merck
All Photos(1)

Documents

BCR635

Human blood (Pb and Cd)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

clinical testing

format

matrix material

storage temp.

−20°C

Application

Human blood (Pb and Cd) may be used as a reference material for the total analysis of microsamples of human blood to detect the presence of cadmium and lead, using high-temperature torch integrated sample introduction system (TISIS) with inductively coupled plasma mass spectrometry(ICP-MS).

Analysis Note

For more information please see:
BCR635

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Total introduction of microsamples in inductively coupled plasma mass spectrometry by high-temperature evaporation chamber with a sheathing gas stream
Grotti M, et al.
Analytica Chimica Acta, 767, 14-20 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service