Skip to Content
Merck
All Photos(1)

Key Documents

BCR276

Zircaloy (C, N, O)

BCR®, certified reference material, cylinder (small, 0.2 g)

Synonym(s):

Zirconium tin alloy

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
€326.00

€326.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 G
€326.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€326.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

form

cylinder (small, 0.2 g)

manufacturer/tradename

JRC

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR276

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service