Skip to Content
Merck
All Photos(1)

Key Documents

93883

Supelco

Fast Yellow AB

analytical standard

Synonym(s):

4-Amino-1,1′-azobenzene-3,4′-disulfonic acid sodium salt, Acid Yellow 9

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H11N3O6S2xNA
CAS Number:
UNSPSC Code:
85151701
E Number:
E105
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

InChI

1S/C12H11N3O6S2.2Na/c13-11-6-3-9(7-12(11)23(19,20)21)15-14-8-1-4-10(5-2-8)22(16,17)18;;/h1-7H,13H2,(H,16,17,18)(H,19,20,21);;/q;2*+1/p-2

InChI key

FPVGTPBMTFTMRT-UHFFFAOYSA-L

General description

Fast Yellow AB (E-105) is used for the quality control in food products. It belongs to the class of water-soluble[1] food color additives. These synthetic food colors are commonly used for the maintenance and improvement of color appearance in foods.[2] Fast Yellow AB is one of the most recalcitrant of the azo dyes due to the arrangement of withdrawing substituents.[3]

Application

Fast Yellow AB may be used as an analytical reference standard for the quantification of the analyte in:
  • Drinks and candies using reversed-phase high-performance liquid chromatography with photodiode array detection.[2]
  • Commercial products such as sports drinks and bakery ink formulations using high-performance thin layer chromatography.[1]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Oxidative decolorization of acid azo dyes by a Mn oxide containing waste
Clarke EC, et al.
Tropical Journal of Pharmaceutical Research, 44(3), 1116-1122 (2010)
Rapid planar chromatographic analysis of 25 water-soluble dyes used as food additives
Morlock EG and Oellig C
Journal of AOAC (Association of Official Analytical Chemists) International, 92(3), 745-756 (2009)
Determination of 40 synthetic food colors in drinks and candies by high-performance liquid chromatography using a short column with photodiode array detection
Yoshioka N and Ichihashi K
Talanta, 764(5), 1408-1413 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service