Skip to Content
Merck
All Photos(1)

Key Documents

936766

Sigma-Aldrich

Josiphos SL-J009-1 Pd G3 ChemBeads

Synonym(s):

Josiphos Pd G3, Josiphos SL-J009-1-G3-palladacycle, {(R)-1-[(Sp)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-tert-butylphosphine}[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

936766-1G
€125.00

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
936766-1G
€125.00

About This Item

Empirical Formula (Hill Notation):
C45H65FeNO3P2PdS
CAS Number:
Molecular Weight:
924.28
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

composition

, 5 wt. % (loading of catalyst)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

NC1=C(C=CC=C1)C2=C([Pd]OS(C)(=O)=O)C=CC=C2.C[C@@H](P(C(C)(C)C)C(C)(C)C)[C]3[C](P(C4CCCCC4)C5CCCCC5)[C][C][C]3.[C]6[C][C][C][C]6.[Fe]

InChI

1S/C27H47P2.C12H10N.C5H5.CH4O3S.Fe.Pd/c1-21(29(26(2,3)4)27(5,6)7)24-19-14-20-25(24)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-2-4-5-3-1;1-5(2,3)4;;/h14,19-23H,8-13,15-18H2,1-7H3;1-6,8-9H,13H2;1-5H;1H3,(H,2,3,4);;/q;;;;;+1/p-1/t21-;;;;;/m1...../s1

InChI key

GJFJVCUFUWXFEB-RPDSBUJNSA-M

General description

Josiphos SL-J009-1 Pd G3 (Josiphos SL-J009-1-G3-palladacycle) is a modified G3 precatalyst (G3′). It has been synthesized by Buchwald and coworkers from second generation (G2) precatalyst by methylation of the amino group present on biphenyl backbone. It is a useful catalyst for various cross-coupling reactions. G3 precatalysts are versatile catalysts. They have long life in solution and are readily soluble in various organic solvents. G3- precatalysts have been employed in various C-N bond forming reactions.[1]

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For larger scale uses, product also available in powdered form 747130

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service