Skip to Content
Merck
All Photos(1)

Key Documents

92255

Supelco

δ-Dodecalactone

analytical standard

Synonym(s):

(±)-5-Dodecanolide, (±)-δ-Heptyl-δ-valerolactone, (±)-6-Heptyltetrahydro-2H-pyran-2-one, 5-Hydroxydodecanoic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
Beilstein:
1282749
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.458-1.462
n20/D 1.460 (lit.)

bp

140-141 °C/1 mmHg (lit.)

mp

−12 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCC1CCCC(=O)O1

InChI

1S/C12H22O2/c1-2-3-4-5-6-8-11-9-7-10-12(13)14-11/h11H,2-10H2,1H3

InChI key

QRPLZGZHJABGRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

δ-Dodecalactone may be used as an analytical reference standard for the quantification of the analyte in wine samples using gas chromatography with selective tandem mass spectrometric detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative analysis of ??and ??lactones in wines using gas chromatography with selective tandem mass spectrometric detection
Langen J, et al.
Rapid Communications in Mass Spectrometry, 27(24), 2751-2759 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service