Skip to Content
Merck
All Photos(1)

Key Documents

91032

Supelco

Ferrocenoyl chloride

for HPLC derivatization, LiChropur, ≥96.0% (AT)

Synonym(s):

1-Chlorocarbonylferrocene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€406.00

€406.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€406.00

About This Item

Empirical Formula (Hill Notation):
C11H9ClFeO
CAS Number:
Molecular Weight:
248.49
MDL number:
UNSPSC Code:
12000000
NACRES:
NA.22

€406.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥96.0% (AT)

form

crystals

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.ClC(=O)[C]2[CH][CH][CH][CH]2

Application

Derivatization reagent for phenols and alcohols[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of trace amounts of alcohols and phenols in complex mixtures as ferrocenecarboxylic acid esters with gas chromatography-atomic emission detection.
Rolfes, J. and Andersson, J.T.
Analytical Communications, 33, 429-432 (1996)
Wenhao Qian et al.
Nanomaterials (Basel, Switzerland), 9(2) (2019-03-03)
A facile strategy to prepare GO-based nanocomposites with both gold nanoparticles (AuNPs) and ferrocene (Fc) moieties was developed. The surface of GO was modified with PFcMAss homopolymer by surface-initiated atom transfer radical polymerization of a new methacrylate monomer of 2-((2-(methacryloyloxy)ethyl)disulfanyl)ethyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service