Skip to Content
Merck
All Photos(1)

Key Documents

90025

Supelco

α,4-Dimethylphenylacetic acid

pharmaceutical impurity standard

Synonym(s):

(2RS)-2-(4-Methylphenyl)propionic acid, Ibuprofen impurity D (PhEur), 2-(4-Methylphenyl)propanoic acid, p-Methylhydratropic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
164.20
Beilstein:
2251315
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

API family

ibuprofen

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

37-42 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C(O)=O)c1ccc(C)cc1

InChI

1S/C10H12O2/c1-7-3-5-9(6-4-7)8(2)10(11)12/h3-6,8H,1-2H3,(H,11,12)

InChI key

KDYOFXPLHVSIHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service