Skip to Content
Merck
All Photos(1)

Documents

89898

Supelco

Acetamide

analytical standard

Synonym(s):

Amide C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH2
CAS Number:
Molecular Weight:
59.07
Beilstein:
1071207
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 65 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

221 °C (lit.)

mp

78-80 °C (lit.)
78-82 °C

format

neat

SMILES string

CC(N)=O

InChI

1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)

InChI key

DLFVBJFMPXGRIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sommer et al.
The international journal of neuropsychopharmacology, 17(12), 2045-2056 (2014-06-26)
Drugs that are able to shift effort-related decision making in intact rats towards high-effort response options are largely unknown. Here, we examined the effects of two candidate drugs, MRZ-9547 and its l-enantiomer MRZ-9546 on progressive ratio (PR) responding using two
Acetamide.
IARC monographs on the evaluation of carcinogenic risks to humans, 71 Pt 3, 1211-1221 (1999-09-07)
Lisa Elviri et al.
Rapid communications in mass spectrometry : RCM, 28(13), 1544-1552 (2014-05-28)
The control of drug release involves gaining an understanding of the complex interaction networks among drug-excipients-matrix-biological fluids. Thus, novel analytical methods that will lead to a better understanding of these interaction networks are urgently required. Desorption electrospray ionization high-resolution mass
I Drira et al.
Mycoses, 57(8), 453-459 (2014-03-14)
The Trichophyton mentagrophytes complex is the main cause of superficial mycoses in humans and animals. Molecular research has provided useful insights into the taxonomy of this complex to overcome the challenges with conventional diagnostics. The aim of this study was
Joana F Sacramento et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 70, 107-116 (2015-02-11)
Caffeine, a non-selective adenosine antagonist, has distinct effects on insulin sensitivity when applied acutely or chronically. Herein, we investigated the involvement of adenosine receptors on insulin resistance induced by single-dose caffeine administration. Additionally, the mechanism behind adenosine receptor-mediated caffeine effects

Articles

A shortened review of fundamentals of Porous Graphitic Carbon (PGC) materials. What they are and how do they work as a stationary phase material in HPLC?.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service